Learn More
Olympus™ Graticule
Product Code. 12502357
Click to view available options
Type:
Line Eyepiece Micrometer
Unit Size:
Each
Product Code. 12502357
Brand: Olympus™ K10017195
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Make accurate measurements of specimens with this eyepiece micrometer. Olympus™ Graticule is a line-type micrometer glass-engraved with a 10mm line with 0.1mm divisions.
  • Glass-engraved.
  • Line-type.
Specifications
Type Line Eyepiece Micrometer
Description 10mm line, 0.1mm divisions on X-axis
For Use With (Equipment) WHN10X, WHN10X-H, WH10X, WH10X-H, WH15X, WHS10X-H, WHS15X-H, CWH10X, CWH10X-H