Learn More
Olympus™ Graticule
Click to view available options
Type:
Line Eyepiece Micrometer
Unit Size:
Each
Description
- Glass-engraved.
- Line-type.
Specifications
Specifications
| Type | Line Eyepiece Micrometer |
| Description | 10mm line, 0.1mm divisions on X-axis |
| For Use With (Equipment) | WHN10X, WHN10X-H, WH10X, WH10X-H, WH15X, WHS10X-H, WHS15X-H, CWH10X, CWH10X-H |