Learn More
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Description
Graphite plate is used for fuel cells, electrolysis, electroplating, and other various mechanical applications where friction, heat, or electrical properties are factors. It is also used as a solid plate for SEM calibration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Graphite plate, pyrolytic |
| Name Note | 1.27x9.98x9.98mm (0.05x0.393x.393 in.) |
| MDL Number | MFCD00144065 |
| Quantity | 1 Pc. |
| Solubility Information | Insoluble in water. |