Learn More
Thermo Scientific Chemicals Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Product Code. 15426055
Click to view available options
Capacity (Metric):
7.5 cc
7.88 cc
Quantity:
36 Pieces
50 Pieces
Unit Size:
Pack of 36
Pack of 50
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15426055
Brand: Thermo Scientific Chemicals 014747.UB
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Unpurified extruded graphite
Material Graphite
For Use With (Application) Metallurgy
Quantity 36 Pieces
Capacity (Metric) 7.88 cc
Product Type Graphite fusion crucible
Shape Drillpoint