Learn More
Trajan Scientific and Medical™ Graphite Ferrules for Shimadzu
Provide leak-free system connections, leading to improved chromatography for non-MS systems.
Brand: Trajan Scientific and Medical™ 0726080
Additional Details : Weight : 0.80000kg
Description
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material, forms a soft grip to the capillary column
- Low emissions
Specifications
GC14 A, GC17 A, GC2010 and GC2014 Detector / Injectors | |
Graphite | |
Graphite Ferrule for Shimadzu 0.1-0.32mm ID Column | |
0.10 to 0.32 mm |