Learn More
asecos™ Granulate Universal Ultra Fine Grain
Product Code. p-200003739 Shop All Asecos Products
Click to view available options
Weight (Metric):
10 kg
30 kg
Unit Size:
Each
Pack of 3
Product Code. 17866566
Brand: asecos™ 14394
This item requires a price update prior to being ordered.
Request a Price for this item here.
Granulate Universal Ultra Fine Grain is ideal for setting oils, alkalis, lipids, fuels, lacquers etc.
  • 0.3-0.7 mm grain size
  • 10 kg bag binds up to 11 l heating oil
  • Penetrates the finest recesses and cracks
  • Ideally suited for applications where little waste and very high suction power is required
  • Available in one or three 10 kg bags.
Specifications
Material Loose Particulate
Type Absorbent Granules
Weight (English) 22.05 lb.
Weight (Metric) 10 kg