Learn More
Grant Instruments™ Grant Plastic Bath Tank
Product Code. p-8014618
Click to view available options
Capacity (Metric):
5 L
12 L
18 L
Dimensions (L x W x H):
390 x 200 x 380 mm
415 x 350 x 380 mm
600 x 350 x 380 mm
Unit Size:
Each
Product Code. 11486437
Brand: Grant Instruments™ P12
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Withstands temperatures up to 99°C.
  • Can be combined for use with any Grant digital thermostat or heated circulating bath
  • Plastic construction
  • 85/140mm min/max liquid depth
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 12 L
Temperature Range 99°C to 99°C
Dimensions (L x W x H) 415 x 350 x 380 mm