Learn More
Hecht Karl™ Graduated Glass Cylinder
Click to view available options
Capacity (Metric):
10 mL
100 mL
200 mL
250 mL
1000 mL
Height (Metric):
90 mm
195 mm
250 mm
265 mm
325 mm
440 mm
Description:
Low Form
Tall Form
Unit Size:
Each
Specifications
Specifications
| Description | Tall Form |
| Height (Metric) | 265 mm |
| Type | Graduated Cylinder |
| Material | Glass |
| Capacity (Metric) | 200 mL |