Learn More
CBS Scientific Gradient Markers
Product Code. p-4085751
Click to view available options
Volume (Metric):
50 mL
250 mL
Unit Size:
Each
Product Code. 11789545
Brand: CBS Scientific GM100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Linear with side outlets.
  • Gradient flow is controlled by a PTFE-coated valve centered between the two chambers
  • The outflow channel is fitted with a male luer adapter which can be used with any needle or luer valve
Specifications
For Use With (Application) Electrophoresis
Type Gradient Marker
Volume (English) 1.69 oz.
Volume (Metric) 50 mL