Learn More
Fisherbrand™ Grade 925 Sterilization Paper
Product Code. 11764576 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
35 cm
50 cm
60 cm
65 cm
75 cm
90 cm
100 cm
Unit Size:
Pack of 2000
Pack of 250
Pack of 500
Product Code. 11764576
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Used as wrap for medical clothes.
  • Creped paper
  • Weight: 60g/mm2
Applications:
  • Used in steam gas and gamma-ray sterilization
Specifications
Autoclavable Autoclavable
Certifications/Compliance EN 868-2
Color White
For Use With (Application) Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
Material Paper
Quantity 250 Pk.
Length (Metric) 75 cm
Width (Metric) 75 cm
Weight (Metric) 60 gsm
Product Type Sterilization Paper
Grade 925