Learn More
Fisherbrand™ Grade 605 Very Absorbent Joseph Paper
Product Code. 11735375 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
120 x 120 mm
150 x 150 mm
220 x 400 mm
500 x 350 mm
Format:
Folded Sheet
Sheet
Packaging Quantity:
100 Pk.
200 Pk.
50 Pk.
500 Pk.
Unit Size:
Pack of 100
Pack of 200
Pack of 50
Pack of 500
Product Code. 11735375
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Used in laboratories and hospitals. Fisherbrand™ Grade 605 Very Absorbent Joseph Paper is thin and not fluffy.
Applications:
  • Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles
Specifications
Length (Metric) 500 mm
Width (Metric) 350 mm
Dimensions (L x W) 500 x 350 mm
Format Folded Sheet
For Use With (Application) Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals
Packaging Quantity 100 Pk.
Grade 605