Learn More
Sartorius™ Grade 470 Folded Filter Cellulose and Diatomaceous Earth Filter Papers
Product Code. 10002291 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
125 mm
150 mm
185 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10002291 150 mm Pack of 100
10270052 125 mm Pack of 100
10286131 185 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10002291 Supplier Sartorius™ Supplier No. FT4606150
 Restricted Item
Estimated Shipment: 29-10-2026
Offers better separating capability and quickly retains the finest particles at high flow rates.
  • Used for the filtration of the finest, semi-colloidal precipitates, e.g. those consisting of proteins, clay or cold-precipitated barium
  • For clarification of beer, wine, urine during spectophotometric or refractometric test
TRUSTED_SUSTAINABILITY
Specifications
Format Folded
Material Cellulose, Diatomaceous Earth
Filtration Speed 80sec.
For Use With (Application) Offer a much better separating capability than pure cellulose papers at the same rate of filtration
Grade 470
Quantity 100/Pk.
Thickness (Metric) 0.32 mm
Type Qualitative Filter Paper
Diameter (Metric) 150 mm