Learn More
Sartorius Stedim Biotech™ Grade 389 Quantitative Papers, Discs
Shop All Sartorius Products
Click to view available options
Diameter (Metric):
600 mm
Unit Size:
Pack of 100
Description
- Made of 100% cotton linters with > 98% alpha-cellulose
- Acid washed to make the papers ashless and achieve high purity (ash content ≥ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
Specifications
| Basis Weight | 84 g/m2 |
| Format | Disc |
| Material | 100% Cotton Linters (>98%alpha-cellulose, Ash-free) |
| Particle Retention | 8 to 12 μm |
| Filtration Speed | 20 sec. |
| Grade | 389 |
| Quantity | 100/Pk. |
| Thickness (Metric) | 0.19 mm |
| Diameter (Metric) | 600 mm |