Learn More
Cytiva Grade 3014 Seed Test Paper
Product Code. 11308904
Click to view available options
Dimensions (L x W):
110 x 20 mm
Unit Size:
Pack of 1000
Product Code. 11308904
Brand: Cytiva 10344676
For testing particularly sensitive seeds. Whatman™ Grade 3014 Seed Test Paper is a white, pleated strip with wrap strip, 50 double pleats made of extremely pure cellulose.
Owing to their extremely high purity our seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances that could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.
TRUSTED_SUSTAINABILITY
Specifications
Basis Weight 113 g/m2
Format Pleated Strip
Material Cellulose
For Use With (Application) Testing particularly sensitive seeds, Medium-large and coated seeds (PP method)
Grade 3014
Includes Wrap Strips
Quantity 100 Pk.
Type Seed Test Paper
Dimensions (L x W) 110 x 20 mm