Learn More
Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 1mm
Product Code. 11336029
Click to view available options
Quantity:
100 Pieces
25 Pieces
5 Pieces
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Product Code. 11336029
Brand: Thermo Scientific™ 042075.KN
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ruby Sphere
Diameter (Metric) 1 mm
Grade Grade 25
Quantity 100 Pieces