Learn More
Munktell Filtrak™ Grade 1600 Pulp Testing Papers
Product Code. 11317254
Click to view available options
Diameter (Metric):
125 mm
140 mm
220 mm
Unit Size:
Pack of 200
Product Code. 11317254
Brand: Munktell Filtrak™ 143039
Only null left
Add to basket
Add to basket
Grade 1600 Test Paper is often used when preparing laboratory sheets of most forms of pulp for physical testing in accordance with ISO Test Method 5269-1: 2005, Part 1.
  • In circle disc format
  • Available in a range of diameters
  • Klemm absorbency of 70 ± 20 mm
  • Dimensional change of less than 3%
  • Water uptake of 450 ± 50 g/m²
  • Provided 200 per pack
TRUSTED_SUSTAINABILITY
Specifications
Basis Weight 250 ± 25 g/m2
Format Disc
For Use With (Application) Physical test prep
Grade Grade 1600
Quantity 200/Pk.
Type Pulp Test Paper
Diameter (Metric) 140 mm