Learn More
Novus Biologicals™ GPSM1 293T Cell Transient Overexpression Lysate
Product Code. 18330989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330989 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18330989 Supplier Novus Biologicals™ Supplier No. H00026086T01
Overexpression Lysate
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction
Target GPSM1
Synonym 1810037C22Rik, C. elegans), DKFZp727I051, G-protein signaling modulator 1, G-protein-signaling modulator 1
Gene Symbol GPSM1