missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GPS1 (Human) Recombinant Protein
Human GPS1 full-length ORF ( AAH00155, 1 a.a. - 475 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002873-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MQIDVDPQEDPQNAPDVNYVVENPSLDLEQYAASYSGLMRIERLQFIADHCPTLRVEALKMALSFVQRTFNVDMYEEIHRKLSEATRSSLRELQNAPDAIPESGVEPPALDTAWVEATRKKALLKLEKLDTDLKNYKGNSIKESIRRGHDDLGDHYLDCGDLSNALKCYSRARDYCTSAKHVINMCLNVIKVSVYLQNWSHVLSYVSKAESTPEIAEQRGERDSQTQAILTKLKCAAGLAELAARKYKQAAKCLLLASFDHCDFPELLSPSNVAIYGGLCALATFDRQELQRNVISSSSFKLFLELEPQVRDIIFKFYESKYASCLKMLDEMKDNLLLDMYLAPHVRTLYTQIRNRALIQYFSPYVSADMHRMAAAFNTTVAALEDELTQLILEGLISARVDSHSKILYARDVDQRSTTFEKSLLMGKEFQRRAKAMMLRAAVLRNQIHVKSPPREGSQGELTPANSQSRMSTNM
Specifications
AAH00155 | |
G protein pathway suppressor 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPS1 | |
GST |
2873 | |
Wheat germ expression system | |
25 μg | |
COPS1, CSN1, MGC71287 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |