Learn More
Abnova™ GPCR Signaling to MAPK/ERk Phospho-Specific Array
Product Code. 16052435
Change view
Click to view available options
Quantity:
1 Kit (2 Samples)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16052435 1 Kit (2 Samples) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16052435 Supplier Abnova™ Supplier No. AA0027.1Kit(2Samples)
GPCR Signaling to MAPK/ERk Phospho-Specific Array includes 193 highly specific and well-characterized phosphorylation antibodies related to this signaling pathway.
  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Specifications
Content And Storage 4°C
Detection Method Fluorometric
Quantity 1 Kit (2 Samples)
For Use With (Application) Profiling
Product Type Array/Library