Learn More
Corning™ Sterile Polypropylene Straight Containers with Polyethylene Caps
Product Code. 12602145
Click to view available options
Capacity (Metric):
60 mL
125 mL
180 mL
Unit Size:
Pack of 264
Pack of 380
Pack of 700
Product Code. 12602145
Brand: Corning™ TP35A009
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Quality sampling tools for secure sample retrieval and shipment
  • Injection molded polypropylene
  • Leakproof polyethylene screw cap
  • Assured Sterile 10-3
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 60 mL
Capacity (English) 2.02 oz.
Color Clear
Material Polypropylene
Sterility Sterile
Closure Color Red
Height (Metric) 70 mm
Closure Material Polyethylene
Diameter (Metric) 33 mm
Closure Type Leakproof Screw Cap