Learn More
Gosselin™ Screw Cap
Product Code. 15921801 Shop All Corning Products
Click to view available options
Color:
Red
Unit Size:
Pack of 3150
Product Code. 15921801
Brand: Gosselin™ C3002
Only null left
Add to basket
Add to basket
Corning™ screw caps have a rapid screw thread, which guarantees complete leak tightness and retains the cap during all handling: transport, centrifugation and storage.
  • Helps to prevent contact with the contents of the tube when screwing and unscrewing
  • Helps to prevent micro-projections when opening the tube.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyethylene
Color Red
No. per Pack 450/Pk.
No. per Case 3150/Cs.