Learn More
Gosselin™ Polystyrene Sampling Spatula
Click to view available options
Length (Metric):
63 mm
85 mm
Unit Size:
Pack of 2000
Pack of 500
Description
Packed in tamper-evident resealable bag
- Polystyrene (PS)
- Packed in tamper-evident resealable bag
Specifications
Specifications
| Sterility | Sterile |
| Color | White |
| Product Type | Sampling Spatula |
| Includes | Peel away certificate |
| Material | Polystyrene |
| Length (Metric) | 63 mm |