Learn More
Gosselin™ Round-Base Test Tubes
Product Code. p-8002486
Click to view available options
Capacity (Metric):
1 mL
2 mL
Closure Type:
Not Included
Straight Cap
Unit Size:
Pack of 2500
Pack of 5000
Product Code. 11700485
Brand: Gosselin™ TL41A03
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Providing high resistance to centrifugation up to 3000g with a suitable adaptor in a balanced rotor. Gosselin™ Round-Base Test Tubes are manufactured from polystyrene plastic.
  • For laboratory use
  • Compatible with automation systems
  • All tubes are CE + IVD marked
  • Available with or without closure
  • Available as either aseptic or sterile
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Red
Closure Material LDPE
Color Natural
Capacity (Metric) 1 mL
Material Polystyrene
For Use With For clinical, dairy, environment, industries, microbiology
Packaging 2500BG
Quantity 10,000 Carton
Rimmed No
Sterility Sterile
Diameter (Metric) 8.4 mm
Closure Type Straight Cap
Product Type Test Tube