Learn More
Motic™ Bifurcated Gooseneck Light Guide
Product Code. 11786576
Change view
Click to view available options
Bulb Type:
Halogen
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bulb Type Unit Size
11786576 Halogen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11786576 Supplier Motic™ Supplier No. 1101002000142
  • ONLINE EXCLUSIVE PRICE
    207.00€
    199.00€ / Pack of 1
    Save 8.00€
    3% Off
Estimated Shipment: 31-12-2026
Requires
Motic MLC-150 cold light illuminator
Half-meter long light guide allows a user to direct light source where it's needed. Motic™ Bifurcated Gooseneck Light Guide is for use with the Motic MLC-150 illuminator.
  • Bifurcated gooseneck style light guide is positionable, holding the angle of illumination required for observations
  • Rugged fiber optic light guide provides cool and uniform illumination, and comes with a PVC coated, stainless steel sheathing
  • 0.5 Meter length
  • Use with MLC-150 halogen cold light illuminator
Specifications
Type Bifurcated Gooseneck
Bulb Type Halogen
For Use With (Equipment) Motic MLC-150 cold light illuminator (11-776-576)
Length (Metric) Arm Reach 0.5 m