missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG Fluor-Linked Secondary Antibody Conjugate, Cytiva
Shop All Cytiva Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
- Species specific antibody supplied in 10mg/mL BSA
- Lyophilised solid in phosphate-buffered saline pH 7.2, containing 1%(w/v) bovine serum albumin and 0.01%(w/v) sodium azide
- Source: Goat
- Immunogen: Mouse IgG, whole molecule
- Gives quantitative results
- Color: White
Applications:
Immunodetection, in situ hybridization, cytometry
Specifications
Specifications
| Description | The antibodies are supplied with dilution guidelines for relevant applications. |
| Concentration | 1 mg/mL |
| For Use With (Application) | Optimal dilutions will depend on the application. Starting dilutions of 1:1000 for microscopy and 1:400 for flow cytometry are recommended |
| Quantity | 1 mg |
| Product Type | Mouse IgG, Cy2-linked (from goat) |
| Purity | This product was prepared from monospecific antiserum by immunoaffinity chromatography using mouse IgG immobilised to agarose beads. This was followed by multiple solid phase adsorptions to eliminate cross reactivity. A single precipitin arc was observed |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C |
| Label or Dye | Cy2 |
| Color | White |
Safety and Handling
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction