missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GNG3 (Human) Recombinant Protein
Human GNG3 full-length ORF ( AAH15563, 1 a.a. - 75 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002785-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKGETPVNSTMSIGQARKMVEQLKIEASLCRIKVSKAAADLVTYCDAHACEDPLITPVPTSENPFREKKFFCALL
Specifications
AAH15563 | |
guanine nucleotide binding protein (G protein), gamma 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2785 | |
Wheat germ expression system | |
25 μg | |
GNG3 | |
GST |