missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GNAI2 (Human) Recombinant Protein
Human GNAI2 full-length ORF ( AAH12138, 1 a.a. - 355 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002771-P02.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGCTVSAEDKAAAERSKMIDKNLREDGEKAAREVKLLLLGAGESGKSTIVKQMKIIHEDGYSEEECRQYRAVVYSNTIQSIMAIVKAMGNLQIDFADPSRADDARQLFALSCTAEEQGVLPDDLSGVIRRLWADHGVQACFGRSREYQLNDSAAYYLNDLERIAQSDYIPTQQDVLRTRVKTTGIVETHFTFKDLHFKMFDVGGQRSERKKWIHCFEGVTAIIFCVALSAYDLVLAEDEEMNRMHESMKLFDSICNNKWFTDTSIILFLNKKDLFEEKITHSPLTICFPEYTGANKYDEAASYIQSKFEDLNKRKDTKEIYTHFTCATDTKNVQFVFDAVTDVIIKNNLKDCGLF
Specifications
AAH12138 | |
guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNAI2 | |
GST |
2771 | |
Wheat germ expression system | |
25 μg | |
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |