Learn More
Novacyt - Microgen Bioproducts GN-ID A 12 Substrate ID System
Product Code. 17383211 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
60 A Test strips
Unit Size:
60 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17383211 60 A Test strips 60 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17383211 Supplier Novacyt - Microgen Bioproducts Supplier No. MID64
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Microgen Bioproducts GN-ID A - For the identification of aerobic and facultative anaerobic gram negative bacteria.
  • Microgen GN-ID A is a convenient 12 substrate ID system for the most commonly isolated Enterobacteriaceae.
  • The Microgen GN-ID A System is for the identification of the commonly encountered Enterobacteriaceae from food or medical samples including urinary samples and common wound isolates.
  • For wider clinical applications and for veterinary and industrial samples, identification of all currently recognised species of Enterobacteriaceae and an extensive range of oxidase-positive Gram Negative Bacilli is achieved with the addition of the GN-ID Bidentification system.
Specifications
Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Gelatin, Malonate, Inositol, Sorbitol, Rhamnose, Sucrose, Lactose, Arabinose, Adonitol, Raffinose, Salicin, Arginine, TDA, Nitrate
Format 60 A Test strips
Sample Type Bacterial Isolate
Sensitivity >99%
Type Test Strip
For Use With (Application) An Identification System for All Currently Recognised Enterobacteriaceae and an Extensive Range of Oxidase-Positive Gramnegative Bacilli
Includes 60 A Test strips
No. of Reactions 12/Test Strip
Organism Group Monera
Packaging Type Carton with pouches and strip holding tray
Quantity 60 A Test strips
Species All currently recognised Enterobacteriaceae and an extensive range of oxidase-positive Gramnegative Bacilli
Specificity >99%
Target Organism Class Bacteria
Test Time 18 to 24 hours incubation for Enterobacteriaceae, and after 48 hours for oxidase positive isolates
Mehr anzeigen Weniger anzeigen