Learn More
Glycylglycine, ≥98.5%
Product Code. 15835158 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835158 100 g Plastic Bottle 100g
15855298 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15835158 Supplier Thermo Scientific Supplier No. J16505.22
Request Bulk or Custom Format
CAS: 556-50-3 | C4H8N2O3 | 132.119 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N
Specifications
Melting Point ∼220°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2CH2CONHCH2COOH
Beilstein 1765223
Quantity 100 g
Formula Weight 132.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Glycylglycine
compliance-icons
Product Identifier
  • Glycylglycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Glycylglycine