missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycoursodeoxycholic-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Glycoursodeoxycholic Acid-d4 |
| CAS | 2044276-17-5 |
| Molecular Formula | C26 D4 H39 N O5 |
| Quantity | 0.01 g |
| Synonym | Glycine, N-[(3α,5β,7β)-3,7-dihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]-, Glycoursodeoxycholic-2,2,4,4-D4 acid, Glycoursodeoxycholic acid-d4, N-[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]glycine, 3α,7β-Dihydroxy-5β-cholanoylglycine-d4, Glycylursodeoxycholic-2,2,4,4-d4 acid, N-(3α,7β-Dihydroxy-5β-cholan-2,2,4,4-d4-24-oyl)glycine, Ursodeoxycholylglycine-d4 |
| InChI Formula | InChI=1 S/C26H43NO5/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29/h15-21,24,28-29 H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16+,17-,18-,19+,20+,21+,24+,25+,26-/m1/s1/i8D2,12D2 |
| SMILES | [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 [C@@H](O)C[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)NCC(=O)O |
| IUPAC Name | 2-[[(4 R)-4-[(3 R,5 R,7 S,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3,7-dihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 453.648 |
| Formula Weight | 453.339 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction