Learn More
Glycohyodeoxycholic Acid, TRC
Product Code. 17979524
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979524 10 mg 10mg
17957324 50 mg 50mg
17998874 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17979524 Supplier Toronto Research Chemicals Supplier No. TRCG64139010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13042-33-6
Molecular Formula C26 H43 N O5
Molecular Weight (g/mol) 449.62
Synonym N-[(3α,5β,6α)-3,6-Dihydroxy-24-oxocholan-24-yl]glycine, N-(3α,6α-Dihydroxy-5β-cholan-24-oyl)glycine, Glycine Hyodeoxycholate, Glycylhyodeoxycholic Acid;
IUPAC Name 2-[[(4R)-4-[(3R,5R,6S,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
SMILES C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C26H43NO5/c1-15(4-7-23(30)27-14-24(31)32)18-5-6-19-17-13-22(29)21-12-16(28)8-10-26(21,3)20(17)9-11-25(18,19)2/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16-,17+,18-,19+,20+,21+,22+,25-,26-/m1/s1
Formula Weight 449.3141
Percent Purity >95%
Chemical Name or Material Glycohyodeoxycholic Acid
Grade HPLC