missing translation for 'onlineSavingsMsg'
Learn More

Merck Glycocholic Acid, Sodium Salt, 100.1%, Calbiochem™,

Product Code. 17866554
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
17866554 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17866554 Supplier Merck Supplier No. 3605125GM
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 863-57-0
Molecular Formula C26H42NNaO6
Molecular Weight (g/mol) 487.61
MDL Number MFCD00036741
InChI Key OABYVIYXWMZFFJ-XIMBWSGONA-M
PubChem CID 134129524
IUPAC Name sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate
SMILES [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity 100.1% (Titration)(dry basis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.