Learn More
Glycocholic Acid Hydrate, TRC
Product Code. 30134780
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134780 10 g 10g
30135389 25 g 25g
30004091 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134780 Supplier Toronto Research Chemicals Supplier No. TRCG64135010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1192657-83-2
Molecular Formula C26 H43 N O6 . x[H2 O]
Molecular Weight (g/mol) 465.52
Synonym N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine Hydrate, N-Choloylglycine-d4, Glycoreductodehydrocholic Acid Hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic αcid-24-glycine Hydrate, Cholylglycine Hydrate, Glycine Cholate Hydrate;
IUPAC Name 2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid;hydrate
SMILES O.C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C26H43NO6.H2O/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);1H2/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;/m1./s1
Formula Weight 483.3196
Percent Purity >95%
Chemical Name or Material Glycocholic Acid Hydrate
Grade HPLC