Learn More
Glycocholic Acid Hydrate Sodium Salt, TRC
Product Code. 30006137
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006137 25 g 25g
30009701 50 g 50g
30010903 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006137 Supplier Toronto Research Chemicals Supplier No. TRCG64136025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 338950-81-5
Molecular Formula C26 H42 N O6 . Na . H2 O
Molecular Weight (g/mol) 505.62
Synonym Glycine, N-[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl]-, monosodium salt, hydrate (9CI), Glycocholic acid sodium salt hydrate, Sodium glycocholate hydrate, N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine sodium salt hydrate, Glycocholic acid sodium salt hydrate, Monosodium glycocholate hydrate, Sodium cholylglycinate hydrate
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate;hydrate
SMILES O.[Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Specifications
Quantity 25 g
InChI Formula InChI=1S/C26H43NO6.Na.H2O/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);;1H2/q;+1;/p-1/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;;/m1../s1
Formula Weight 505.302
Chemical Name or Material Glycocholic Acid Hydrate Sodium Salt