missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycocholic-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Glycocholic-2,2,4,4-d4 Acid |
| CAS | 1201918-15-1 |
| Molecular Formula | C26 D4 H39 N O6 |
| Quantity | 0.01 g |
| Synonym | Glycine, N-[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]- (ACI), N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]glycine (ACI), Glycocholic-2,2,4,4-D4 acid, Glycocholic acid-d4, Glycocholic-d4 acid, Cholyl-2,2,4,4-d4-glycine |
| InChI Formula | InChI=1 S/C26H43NO6/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29/h14-21,24,28-30 H,4-13H2,1-3H3,(H,27,31)(H,32,33)/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-/m1/s1/i8D2,10D2 |
| SMILES | [2 H]C1([2 H])C[C@]2(C)[C@H]3 C[C@H](O)[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 [C@H](O)C[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)NCC(=O)O |
| IUPAC Name | 2-[[(4 R)-4-[(3 R,5 R,7 R,8 R,9 S,10 S,12 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3,7,12-trihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 469.6474 |
| Formula Weight | 469.3341 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction