Learn More
Glycitein
Product Code. 16504434 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
Product Code. 16504434
Brand: Thermo Scientific Acros 464640500
Request Bulk or Custom Format
CAS: 40957-83-3 | C16H12O5 | 284.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40957-83-3
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
InChI Key DXYUAIFZCFRPTH-UHFFFAOYSA-N
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one
SMILES COC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1
Specifications
Quantity 50 mg
Formula Weight 284.26
Chemical Name or Material Glycitein
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.