missing translation for 'onlineSavingsMsg'
Learn More

Glycine t-Butyl Ester Hydrochloride, TRC

Product Code. 30402882
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402882 1 g
30405103 5 g
30399748 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402882 Supplier Toronto Research Chemicals Supplier No. TRCG6251751G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6 H13 N O2 . Cl H
Molecular Weight (g/mol) 167.63
IUPAC Name tert-butyl 2-aminoacetate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CN

Specifications

InChI Formula InChI=1S/C6H13NO2.ClH/c1-6(2,3)9-5(8)4-7;/h4,7H2,1-3H3;1H
Quantity 1 g
Formula Weight 167.0713
Shipping Condition Room Temperature
Chemical Name or Material Glycine, t-Butyl Ester, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.