Learn More
Glycine-d5, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Glycine-d5 |
| CAS | 4896-77-9 |
| Molecular Formula | C2 2H5 N O2 |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1D2/hD3 |
| SMILES | [2 H]OC(=O)C([2 H])([2 H])N([2 H])[2 H] |
| IUPAC Name | deuterio 2,2-dideuterio-2-(dideuterioamino)acetate |
| Molecular Weight (g/mol) | 80.1 |
| Formula Weight | 80.0634 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |