Learn More
Glyceryl Iodide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-iodopropane-1,2-diol |
| CAS | 554-10-9 |
| Molecular Formula | C3 H7 I O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C3H7IO2/c4-1-3(6)2-5/h3,5-6H,1-2H2 |
| SMILES | OCC(O)CI |
| IUPAC Name | 3-iodopropane-1,2-diol |
| Molecular Weight (g/mol) | 201.991 |
| Formula Weight | 201.949 |