Learn More
GlycanTune™ A+ Total Feed
Click to view available options
Quantity:
10 L
Unit Size:
10 L
Specifications
Specifications
| Content And Storage | AGT™ dry format. Store in refrigerator (2–8°C). Protect from light and moisture. |
| Product Type | Total Feed |
| Product Line | GlycanTune |
| Quantity | 10 L |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |