Learn More
MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin
Product Code. 11449795 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449795 25mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11449795 Supplier MP Biomedicals™ Supplier No. 03AFC03325
  • ONLINE EXCLUSIVE PRICE
    508.00€
    483.00€ / 25mg
    Save 25.00€
    4% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder
  • Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
  • In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
  • It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
Specifications
Color White
Form Powder
For Use With (Application) Cell Migration Analysis; Cell Signaling Analysis
Quantity 25mg
Molecular Weight (g/mol) 433.38 ; 574 (Including TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Keep cool and dry