missing translation for 'onlineSavingsMsg'
Learn More

Glutaronitrile, 99%

Product Code. 11490068
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11490068

Brand: Thermo Scientific Alfa Aesar B23663.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-13-8 | C5H6N2 | 94.117 g/mol

It can be used for the conversion, by exchange, of carboxylic acids to nitriles, by heating in the presence of phosphoric acid. The equilibrium is displaced by cyclization to glutarimide. It is also used in the preparation of dicarbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used for the conversion, by exchange, of carboxylic acids to nitriles, by heating in the presence of phosphoric acid. The equilibrium is displaced by cyclization to glutarimide. It is also used in the preparation of dicarbonyl compounds.

Solubility
Soluble in water, alcohol and chloroform.

Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-13-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00001970
InChI Key ZTOMUSMDRMJOTH-UHFFFAOYSA-N
Synonym glutaronitrile, 1,3-dicyanopropane, glutarodinitrile, glutaric acid dinitrile, 1,3-trimethylenedinitrile, pyrotartaric acid nitrile, glutaronitrle, unii-01zi68f3cq, trimethylene dicyanide, glutaric dinitrile
PubChem CID 10994
IUPAC Name pentanedinitrile
SMILES C(CC#N)CC#N

Specifications

Melting Point -29°C
Density 0.99
Boiling Point 286°C to 288°C
Flash Point 112°C (234°F)
Refractive Index 1.435
Quantity 50 g
UN Number UN3276
Beilstein 1738385
Merck Index 14,4474
Solubility Information Soluble in water,alcohol and chloroform.
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material Glutaronitrile
Show More Show Less
compliance-icons
Product Identifier
  • Glutaronitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Pentanedinitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.