Learn More
Glucose Pentasulfate, Potassium Salt, Technical Grade, TRC
Specifications
Specifications
| Chemical Name or Material | Glucose Pentasulfate, Potassium Salt, Technical Grade |
| CAS | 359435-44-2 |
| Molecular Formula | C6H7O21S5K5 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H12O21S5.5K/c7-28(8,9)22-1-2-3(24-29(10,11)12)4(25-30(13,14)15)5(26-31(16,17)18)6(23-2)27-32(19,20)21;;;;;/h2-6H,1H2,(H,7,8,9)(H,10,11,12)(H,13,14,15)(H,16,17,18)(H,19,20,21);;;;;/q;5*+1/p-5/t2-,3-,4+,5-,6?;;;;;/m1...../s1 |
| SMILES | [K+].[K+].[K+].[K+].[K+].[O-]S(=O)(=O)OC[C@@H](OS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@H](OS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])C=O |
| Molecular Weight (g/mol) | 770.94 |