missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GLO1 (Human) Recombinant Protein
Human GLO1 full-length ORF ( AAH11365, 1 a.a. - 184 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00002739-P01.25ug
Informations supplémentaires : Weight : 0.00010kg
Description
- Sequence: MAEPQPPSGGLTDEAALSYCSDADPSTKDFLLQQTMLRVKDPKKSLDFYTRVLGMTLIQKCDFPIMKFSLYFLAYEDKNDIPKEKDEKIAWALSRKATLELTHNWGTEDDETQSYHNGNSDPRGFGHIGIAVPDVYSACKRFEELGVKFVKKPDDGKMKGLAFIQDPDGYWIEILNPNKMATLM
Spécification
AAH11365 | |
glyoxalase I | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLO1 | |
GST |
2739 | |
Wheat germ expression system | |
25 μg | |
GLOD1, GLYI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |