Learn More
Glimepiride-d4 (phenylethyl-a,a,b,b-d4) (cis/trans), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (cis/trans)-Glimepiride-D4 (2-phenylethylamine-1,1,2,2-D4) |
| CAS | 2699607-96-8 |
| Molecular Formula | C24 D4 H30 N4 O5 S |
| Quantity | 0.005 g |
| Synonym | Glimepiride-d4 (phenylethyl-a,a,b,b-d4) (cis/trans), 3-Ethyl-2,5-dihydro-4-methyl-N-[2-[4-[[[[(4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl-1,1,2,2-d4]-2-oxo-1 H-pyrrole-1-carboxamide, (cis/trans)-Glimepiride-d4, (cis/trans)-Glimepiride-D4 (2-phenylethylamine-1,1,2,2-D4) |
| InChI Formula | InChI=1 S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19 H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/i13D2,14D2 |
| SMILES | [2 H]C([2 H])(NC(=O)N1CC(=C(CC)C1=O)C)C([2 H])([2 H])c2ccc(cc2)S(=O)(=O)NC(=O)NC3CCC(C)CC3 |
| IUPAC Name | 4-ethyl-3-methyl-5-oxo-N-[1,1,2,2-tetradeuterio-2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-2 H-pyrrole-1-carboxamide |
| Molecular Weight (g/mol) | 494.6402 |
| Formula Weight | 494.2501 g/mol |
| Show More |