Learn More
GLB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA595966
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: TSYDYDAPLS EAGDLTEKYF ALRNIIQKFE KVPEGPIPPS TPKFAYGKVT LEKLKTVGAA LDILCPSGPI KSLYPLTFIQ VKQHYGFVLY RTTLPQDCSN PAPLSSPLNG VHDRAYVAVD GIPQGVLERN NVITLNITGK AGATLDLLVE NMGRVNYGAY INDFKGLVSN LTLSSNILTD WTIFPLDTED AVCSHLGGWG HRDSGHHDEA WAHNSSNYTL PAFYMGNFSI PSGIPDLPQD TFIQFPGWTK GQVWINGFNL GRYWPARGPQ LTLFVPQHIL MTSAPNTITV LELEWAPCSS DDPELCAVTF VDRPVIGSSV TYDHPSKPVE KRLMPPPPQK NKDSWLDHV Positive Samples: A549, HeLa, Mouse kidney; Cellular Location: Cytoplasm, Lysosome, perinuclear region
Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.Specifications
GLB1 | |
Polyclonal | |
Unconjugated | |
GLB1 | |
acid beta-galactosidase, AW125515, betagal, Beta-Gal, Beta-galactosidase, bgal, Bge, Bgl, Bgl-e, Bgl-s, Bgl-t, Bgs, Bgt, C130097A14Rik, EBP, Elastin receptor 1, elastin receptor 1, 67kDa, ELNR1, galactosidase beta 1, galactosidase, beta 1, GLB1, Glb-1, lactase, lysosmal hydrolase, lysosomal beta-galactosidase, lysosomal hydrolase, MPS4B, βgal, β-gal, β-galactosidase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12091, 2720, 316033 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3 | |
P16278, P23780 | |
GLB1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 329-677 of human GLB1 (NP_0003953) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |