Learn More
Thermo Scientific™ Glassy Carbon Tapered Crucible for Crystal Growth
Product Code. 15403755
Click to view available options
Capacity (Metric):
17 mL
18 mL
37 mL
105 mL
190 mL
277 mL
370 mL
Unit Size:
Each
Product Code. 15403755
Brand: Thermo Scientific™ 040956.KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.
Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
TRUSTED_SUSTAINABILITY
Specifications
Description Tapered
Material Glassy Carbon
For Use With (Application) For Crystal Growth
Capacity (Metric) 105 mL
Diameter (Metric) Outer Top: 59mm
Height (Metric) 59mm
Product Type Glassy Carbon Crucible