Learn More
Thermo Scientific Chemicals Glassy Carbon E-Beam Evaporating Crucible
Product Code. 15443755
Click to view available options
Capacity (Metric):
4 mL
7 mL
8 mL
10 mL
16 mL
30 mL
Unit Size:
Each
Product Code. 15443755
Brand: Thermo Scientific Chemicals 040960.KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.
Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
TRUSTED_SUSTAINABILITY
Specifications
Description Angle: 15 deg.
Material Glassy Carbon
For Use With (Application) E-Beam Evaporating
Capacity (Metric) 4 mL
Diameter (Metric) Outer 29mm
Height (Metric) 15mm
Product Type Glassy Carbon Crucible
Diameter (Metric) Base 22mm