Learn More
Thermo Scientific Chemicals Glassy Carbon Boat;Vol (ml), 3;Length (mm), 53;Width (mm), 16;Ht (mm),10
Click to view available options
Dimensions (L x W x H):
29 x 16 x 10 mm
53 x 16 x 10 mm
76 x 33 x 25 mm
103 x 16 x 10 mm
Capacity (Metric):
2 mL
3 mL
10 mL
40 mL
Unit Size:
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Resistance to all wet decomposition agents and abrasive wear
- No memory effects or contamination of analytical samples
- Stability to acid and alkaline melts
- No wetting effect of metal melts
- Resistance to thermal shock allows rapid heating and cooling times
- Good electrical conductivity
- Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal growing such as doped halogenide crystals
Specifications
Specifications
| Type | Glassy Carbon Boat |
| Capacity (Metric) | 40 mL |
| Dimensions (L x W x H) | 76 x 33 x 25 mm |
| Height (Metric) | 25 mm |
| Length (Metric) | 76 mm |
| Width (Metric) | 33 mm |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering