Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10
Product Code. 15717818
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15717818 100 Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15717818 Supplier Kova Plastics by Alltrista Supplier No. 87146E
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis.
  • Made from optically clear plastic
  • Glasstic Slide features 10 individual fully segregated specimen wells
  • Capillary action reduces the likelihood of pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
  • Correlate well with results obtained by using a hemocytometer: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
Type Slide 10
For Use With (Application) Microscopy
Gridded No
Material Plastic with the optical qualities of glass
No. of Wells 10
Quantity 100 Pack