Learn More
Fistreem International™ Glass Water Pipettor Transferpette
Product Code. 13156010
Change view
Click to view available options
Voltage:
240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
13156010 240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13156010 Supplier Fistreem International™ Supplier No. WSC044.MH3.7
  • ONLINE EXCLUSIVE PRICE
    6205.00€
    6068.51€ / Pack of 1
    Save 136.49€
    2% Off
 Restricted Item
Estimated Shipment: 15-01-2027
Cyclon ultrapure water stills are designed to surpass accepted purity and performance levels of traditional systems
Cyclon ultrapure water stills are designed to surpass accepted purity and performance levels of traditional systems. Of particular interest to users in life sciences, the pyrogen free distillate is of the very highest purity. The still's unique condenser vapour trap creates a cyclonic effect eliminating any vapour borne droplets, thereby preventing carry-over in the distillate.Outputs of 4L/hr and 8L/hr single distilled and 4L/hr double distilled are offered. The high quality borosilicate glassware houses very durable, silica sheathed, heating elements. Protection of the heaters by thermistor control and a thermal fuse allows unattended safe operation. Auto reservoir controls automatically switch off the still on a full reservoir. The stills are supplied ready for connection to a mains tap supply and will provide high quality pyrogen free water, even on the hardest feed water supplies. The boiler contents are drained automatically on switch off to reduce the rate of scale formation. The still has a clean facility which simplifies and semi-automates the descaling process.
Treated feed option
Where preferred, Cyclon stills may be fed with a suitable quality pre-treated water to reduce or eliminate the need for periodic cleaning. In this condition the condenser water (tap feed) is separated from the boiler water (treated feed) for economy. Cyclon stills are supplied with pre-treated feed components which are easily fitted by the user to allow treated water to be delivered directly to the boiler. In this condition controls ensure the treated feed water is automatically supplied on demand, so avoiding any wastage. Suitable feed water is deionised, reverse osmosis or distilled quality. The still may be fed from a user's existing water source. Alternatively, the Cyclon deioniser is specially designed for use with the still.
A low water pressure adapter is available for use with feed water pressure between 0.35bar and 0.70bar (5psi to 10psi). Please contact Customer Service for information.
Specifications
Product Type Cyclon Ultrapure Water Still
Electrical Requirements 240 V 6 kW
Voltage 240 V
Depth (Metric) 380 mm
Height (Metric) 750 mm
Width (Metric) 490 mm